Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plfr Antibody (RM0308-7N31), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212437AF405
Description
Plfr Monoclonal specifically detects Plfr in Mouse samples. It is validated for Western Blot.Specifications
Plfr | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rat | |
Protein A or G purified | |
Primary | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0308-7N31 | |
Western Blot | |
PLF-RP, Prl7d1, PRP | |
Mouse recombinant Plfr | |
0.1 mL | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction