Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLGF Antibody (MM0010-2D93), DyLight 755

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060976IR
Description
PLGF Monoclonal specifically detects PLGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PLGF | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PGF | |
Human recombinant PLGF | |
0.1 mL | |
Primary | |
In western blots, this antibody shows no cross-reactivity with human VEGF or PDGF. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0010-2D93 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
Mouse | |
Protein G purified | |
Angiogenesis | |
5228 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction