Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLGF Rabbit anti-Mouse, Clone: 17, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29043050UL
Description
PLGF Monoclonal specifically detects PLGF in Mouse samples. It is validated for ELISA.Specifications
PLGF | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
17 | |
ELISA 1:5000-1:10000 | |
D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
Recombinant Mouse PLGF Protein | |
50 μg | |
Angiogenesis | |
5228 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction