Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLGLA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549130

Catalog No. PA549130


Only null left
Add to Cart

Description

Description

PLGLA Polyclonal Antibody for Western Blot

May bind non-covalently to lysine binding sites present in the kringle structures of plasminogen. This may interfere with the binding of fibrin or alpha-2-antiplasmin to plasminogen and may result in the localization of activity at sites necessary for extracellular matrix destruction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLGLA
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q15195
PLGLA
KLH conjugated synthetic peptide between 23-48 amino acids from the N-terminal region of human PLGLA.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
PLGLA
plasminogen-like A (pseudogene); PLGLA; PLGLA1; PLGP2; PRGA
Rabbit
Antigen affinity chromatography, Protein A
RUO
285189
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.