Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLGLB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565821 Supplier Invitrogen™ Supplier No. PA565821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTCRAFQYHSK EQQCVIMAEN RKSSIIIRMR D Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

May bind noncovalently to lysine binding sites present in the kringle structures of plasminogen. This may interfere with the binding of fibrin or alpha-2-antiplasmin to plasminogen and may result in the localization of activity at sites necessary for extracellular matrix destruction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLGLB1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLGLB1
Gene Accession No. Q02325
Gene Alias plasminogen-like B1; plasminogen-like protein B; Plasminogen-related protein B; PLGL; PLGL PRGB; PLGLB1; PLGLB2; PLGP1; PRGB; PRP-B; type B plasminogen related
Gene Symbols PLGLB1
Host Species Rabbit
Immunogen Recombinant Human PLGLB1. Recombinant protein control fragment (Product #RP-105626).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.