Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLGRKT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107216 Supplier Invitrogen™ Supplier No. PA5107216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 77%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 85%; Rat: 93%.

The exact function of C9orf46 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLGRKT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene PLGRKT
Gene Accession No. Q9HBL7
Gene Alias 1110007H22Rik; 5033414D02Rik; AD025; AI852040; C9orf46; MDS030; Plasminogen receptor (KT); plasminogen receptor with a C-terminal lysine; plasminogen receptor, C-terminal lysine transmembrane protein; Plg-R(KT); PLGRKT; Plg-RKT; RGD1306839; transmembrane protein C9orf46; transmembrane protein C9orf46 homolog
Gene Symbols PLGRKT
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C9orf46 (aa 72-122).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.