Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plicamycin, 1mg/mL in DMSO, sterile-filtered, Thermo Scientific™

Catalog No. AAJ67319XF
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67319XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67319XF Supplier Thermo Scientific Chemicals Supplier No. J67319XF
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18378-89-7
Molecular Formula C52H76O24
Molecular Weight (g/mol) 1085.156
MDL Number MFCD00135618
InChI Key CFCUWKMKBJTWLW-ZUZMCERRSA-N
Synonym Aureolic acid; Mithramycin A
PubChem CID 122172942
IUPAC Name (2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dih
SMILES CC1C(C(CC(O1)OC2CC(OC(C2O)C)OC3=CC4=CC5=C(C(=O)C(C(C5)C(C(=O)C(C(C)O)O)OC)OC6CC(C(C(O6)C)O)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)O)(C)O)C(=C4C(=C3C)O)O)O)O

Specifications

Type Plicamycin
Physical Form Liquid
Quantity 1 mL
Chemical Name or Material Plicamycin, 1mg/mL in DMSO, sterile-filtered
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
RTECSNumber PZ2800000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.