Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLIN3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570162
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0162 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0162 Supplier Bethyl Laboratories, Inc Supplier No. A305336AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

Tail-interacting protein (TIP47) is a cytosolic protein essential for the transport of mannose-6-phosphate receptors (MPRs) from endosomes to the trans-Golgi compartments in cells. TIP47 is recruited from the cytoplasm to the late endosomal surface by the Ras-associated protein Rab9 GTPase, enabling it to bind more efficiently to MPR cytoplasmic domains. Recently, it was shown that TIP47 regulates the expression of Rab9, as expression of siRNA to TIP47 in transfected cells dramatically decreased the half-life of the Rab9 protein in addition to stabilizing the subcellular localization of Rab9. At least two isoforms of TIP47 are known to exist.

Specifications

Antigen PLIN3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PLIN3
Gene Accession No. O60664
Gene Alias M6PRBP1, MGC11117, MGC2012, PP17, TIP47
Gene Symbols PLIN3
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10226
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.