Learn More
PLIN5 Mouse anti-Human, Clone: OTI4C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811642
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Lipid droplet-associated protein that maintains the balance between lipogenesis and lipolysis and also regulates fatty acid oxidation in oxidative tissues. Recruits mitochondria to the surface of lipid droplets and is involved in lipid droplet homeostasis by regulating both the storage of fatty acids in the form of triglycerides and the release of fatty acids for mitochondrial fatty acid oxidation. In lipid droplet triacylglycerol hydrolysis, plays a role as a scaffolding protein for three major key lipolytic players: ABHD5, PNPLA2 and LIPE. Reduces the triacylglycerol hydrolase activity of PNPLA2 by recruiting and sequestering PNPLA2 to lipid droplets. Phosphorylation by PKA enables lipolysis probably by promoting release of ABHD5 from the perilipin scaffold and by facilitating interaction of ABHD5 with PNPLA2. Also increases lipolysis through interaction with LIPE and upon PKA-mediated phosphorylation of LIPE.Specifications
| PLIN5 | |
| Monoclonal | |
| Unconjugated | |
| PLIN5 | |
| LSDA5, LSDP5, MLDP, OXPAT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 440503 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4C8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q00G26 | |
| PLIN5 | |
| Human recombit protein fragment corresponding to amino acids 1-463 of human PLIN5 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.