Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056651T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056651T 0.05 mL
NB10056651 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056651T Supplier Novus Biologicals Supplier No. NB10056651SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

PLK1 Polyclonal specifically detects PLK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PLK1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28474672), Immunohistochemistry-Paraffin
Formulation Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P53350
Gene Alias cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13
Gene Symbols PLK1
Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human PLK1 was used as the immunogen.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5347
Test Specificity Reacts with Bovine, Dog, Human, Mouse, New World monkey, Pig, Rat and Rhesus Monkey. Predicted to react with Zebrafish, C elegans and Xenopus.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.