Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLK1 Antibody (AZ44), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002747IR
Description
PLK1 Monoclonal specifically detects PLK1 in Xenopus samples. It is validated for Western Blot, ELISA.Specifications
PLK1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLK1 | |
Baculo-expressed PLK1 from Xenopus laevis | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
5347 | |
Xenopus species | |
Purified |
Western Blot, ELISA | |
AZ44 | |
Western Blot, ELISA | |
cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Plx is activated by phosphorylation. AZ44 only detects the mitotic form of Plx (Xenopus laevis) which may be due to the recognition of a phosphorylated epitope. Recommended concentration: 1 ug/ml | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction