Learn More
Promega PLK1 Kinase Enzyme System, 10microg
Catalog No. PRV2841 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV2841 10 μg
1 options
Catalog No. PRV2841 Supplier Promega Supplier No. V2841
Recombinant full-length human PLK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. A member of the Polo-Like Kinase family, PLK1 localizes to centrosomes and undergoes dramatic subcellular relocation during the cell cycle.
  • Easily Screen and Profile PLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C