Learn More
Promega PLK1 Kinase Enzyme System, 1mg
Catalog No. PRV2842 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2842 1 mg
1 options
Catalog No. PRV2842 Supplier Promega Supplier No. V2842
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PLK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. A member of the Polo-Like Kinase family, PLK1 localizes to centrosomes and undergoes dramatic subcellular relocation during the cell cycle.
  • Easily Screen and Profile PLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C