Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK1 (PHO T210 Mouse anti-Human, Clone: 2A3, Abnova™

Catalog No. 89934782 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-782 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-782 Supplier Abnova Corporation Supplier No. MAB12911
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of human PLK1.

Specifications

Antigen PLK1 (PHO T210
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 2A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of human PLK1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene Alias PLK/STPK13
Gene Symbols PLK1
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T210 of human PLK1.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5347
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.