Learn More
PLK1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00071T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rabbit, Gorilla, Chimpanzee, Northern white-cheeked gibbon and White-tufted-ear marmoset
Polo-like Kinase 1 (PLK1) is found to have several functions in mitotic progression. It also has been found to be elevated in a broad range of human tumors. The function of PLK1 appears to be a requirement for proper spindle assembly and other centrosome microtubule events.Specifications
PLK1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P53350 | |
PLK1 | |
Between 325 and 375 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PLK1 | |
phospho-polo-like-kinase (Ser482,486,490) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5347 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.