Learn More
PLK1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | PLK1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-7215
|
Bethyl Laboratories, Inc
A300251AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells.
Polo-like Kinase 1 (PLK1) is found to have several functions in mitotic progression. It also has been found to be elevated in a broad range of human tumors. The function of PLK1 appears to be a requirement for proper spindle assembly and other centrosome microtubule events.Specifications
| PLK1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| phospho-polo-like-kinase (Ser482,486,490) | |
| PLK1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P53350 | |
| 5347 | |
| Between 325 and 375 | |
| Primary | |
| 4° C | |
| PLK1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.