Learn More
Invitrogen™ PLK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596158
Description
Immunogen sequence: FFTSGYIPAR LPITCLTIPP RFSIAPSSLD PSNRKPLTVL NKGLENPLPE RPREKEEPVV RETGEVVDCH LSDMLQQLHS VNASKPSERG LVRQEEAEDP ACIPIFWVSK WVDYSDKYGL GYQLCDNSVG VLFNDSTRLI LYNDGDSLQY IERDGTESYL TVSSHPNSLM KKITLLKYFR NYMSEHLLKA GANITPREGD ELARLPYLRT WFRTRSAIIL HLSNGSVQIN FFQDHTKLIL CPLMAAVTYI DEKRDFRTYR LSLLEEYGCC KELASRLRYA RTMVDKLLSS RSASNRLKAS; Positive Samples: HeLa, HepG2, SKOV3, SGC-7901, Mouse testis; Cellular Location: Chromosome, Cytoplasm, Midbody, Nucleus, centromere, centrosome, cytoskeleton, kinetochore, microtubule organizing center, spindle.
PLK1 is a serine/threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis. Polo-like kinase proteins acts by binding and phosphorylating proteins are that already phosphorylated on a specific motif recognized by the POLO box domains. Phosphorylates BORA, BUB1B/BUBR1, CCNB1, CDC25C, CEP55, ECT2, ERCC6L, FBXO5/EMI1, FOXM1, KIF20A/MKLP2, CENPU, NEDD1, NINL, NPM1, NUDC, PKMYT1/MYT1, KIZ, PPP1R12A/MYPT1, PRC1, RACGAP1/CYK4, SGOL1, STAG2/SA2, TEX14, TOPORS, p73/TP73, TPT1 and WEE1. Plays a key role in centrosome functions and the assembly of bipolar spindles by phosphorylating KIZ, NEDD1 and NINL.
Specifications
PLK1 | |
Polyclonal | |
Unconjugated | |
Plk1 | |
cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18817, 25515, 5347 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P53350, Q07832, Q62673 | |
Plk1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 304-603 of human PLK1 (NP_0050212). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.