Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLK1 Rabbit anti-Human, Clone: 8G9X10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $516.00
Specifications
Antigen | PLK1 |
---|---|
Clone | 8G9X10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Human samples. It is validated for Western BlotSpecifications
PLK1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5347 | |
IgG | |
Affinity purified |
8G9X10 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
A synthetic phosphorylated peptide around T210 of human PLK1 (P53350). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title