Learn More
Invitrogen™ PLK2 Polyclonal Antibody

Description
PLK2 Polyclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | PLK2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PLK2 |
| Gene Accession No. | P53351, Q9NYY3, Q9R012 |
| Gene Alias | h PLK-2; hPlk2; hSNK; Plk2; PLK-2; polo like kinase 2; polo-like kinase 1; polo-like kinase 2; Serine/threonine-protein kinase PLK2; serine/threonine-protein kinase SNK; Serum-inducible kinase; SNK |
| Gene Symbols | PLK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.