Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579831
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 cell, human HepG2 cell, mouse heart tissue. IHC: human tonsil tissue .
PLK2 is a serum inducible serine/threonine kinase that is a member of the polo family of serine/threonine kinases which have a role in normal cell division. The kinase may play a role in embryogenesis, wound healing, and neoplasia.
Specifications
PLK2 | |
Polyclonal | |
Unconjugated | |
PLK2 | |
h PLK-2; hPlk2; hSNK; Plk2; PLK-2; polo like kinase 2; polo-like kinase 1; polo-like kinase 2; Serine/threonine-protein kinase PLK2; serine/threonine-protein kinase SNK; Serum-inducible kinase; SNK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10769, 20620, 83722 | |
-20°C | |
Lyophilized |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4 mg trehalose and 0.05 mg sodium azide | |
P53351, Q9NYY3, Q9R012 | |
PLK2 | |
E. coli-derived human PLK2 recombinant protein (Position: A94-Q188). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction