Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579832
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579832 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579832 Supplier Invitrogen™ Supplier No. PA579832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: A431 Cell, 293T Cell, COLO-320 Cell. IHC: human lung cancer tissue, rat intestine tissue, rat brain tissue. ICC/IF: A549 cell. Flow: A549 cell.

PLK2 is a serum inducible serine/threonine kinase that is a member of the polo family of serine/threonine kinases which have a role in normal cell division. The kinase may play a role in embryogenesis, wound healing, and neoplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLK2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene PLK2
Gene Accession No. P53351, Q9NYY3, Q9R012
Gene Alias h PLK-2; hPlk2; hSNK; Plk2; PLK-2; polo like kinase 2; polo-like kinase 1; polo-like kinase 2; Serine/threonine-protein kinase PLK2; serine/threonine-protein kinase SNK; Serum-inducible kinase; SNK
Gene Symbols PLK2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human PLK2 (666-685aa SSELKNRMEYALNMLLQRCN).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10769, 20620, 83722
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.