Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | PLK3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PLK3 Polyclonal specifically detects PLK3 in Mouse samples. It is validated for Western Blot.Specifications
| PLK3 | |
| Polyclonal | |
| Rabbit | |
| CNKpolo-like kinase 3 (Drosophila), cytokine-inducible kinase, Cytokine-inducible serine/threonine-protein kinase, EC 2.7.11, FNKEC 2.7.11.21, polo-like kinase 3PLK-3, PRKFGF-inducible kinase, Proliferation-related kinase, serine/threonine-protein kinase PLK3 | |
| PLK3 | |
| IgG | |
| 55 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 1263 | |
| Synthetic peptides corresponding to the C terminal of Plk3. Immunizing peptide sequence PDVRPEAPVVSGQAPASLVETAAEDSSPRGTLASSGDGFEEGLTVATVVE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title