Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597143 Supplier Invitrogen™ Supplier No. PA597143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLFNDGTHMA LSANRKTVHY NPTSTKHFSF SVGAVPRALQ PQLGILRYFA SYMEQHLMKG GDLPSVEEVE VPAPPLLLQW VKTDQALLML FSDGTVQVNF YGDHTKLILS GWEPLLVTFV ARNRSACTYL ASHLRQLGCS PDLRQRLRYA LRLLRDRSPA; Positive Samples: A-549, HT-1080, SKOV3, 293T, U2OS, Mouse liver, Rat liver; Cellular Location: Cytoplasm, Golgi apparatus, Nucleus, centrosome, cytoskeleton, microtubule organizing center, nucleolus.

PLK3 is a member of the polo family of serine/threonine kinases. PLK3 is involved in the regulation of M phase in the cell cycle. CNK contains both a catalytic domain and a putative regulatory domain. It may play a role in regulation of cell cycle progression and tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLK3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLK3
Gene Accession No. Q60806, Q9H4B4, Q9R011
Gene Alias CNK; cytokine inducible kinase; cytokine-inducible serine/threonine-protein kinase; FGF-inducible kinase; Fnk; PLK3; PLK-3; polo like kinase 3; polo-like kinase 3; PRK; Proliferation-related kinase; serine/threonine-protein kinase PLK3; serine-threonine kinase
Gene Symbols PLK3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 487-646 of human PLK3 (NP_0040642).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1263, 12795, 58936
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.