Learn More
PLK4 Mouse anti-Human, Clone: OTI2D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810507

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the polo family of serine/threonine protein kinases. The protein localizes to centrioles, complex microtubule-based structures found in centrosomes, and regulates centriole duplication during the cell cycle.Specifications
PLK4 | |
Monoclonal | |
Unconjugated | |
PLK4 | |
MCCRP2, SAK, STK18 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10733 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00444 | |
PLK4 | |
Human recombit protein fragment corresponding to amino acids 307-612 of human PLK4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.