Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111050 Supplier Invitrogen™ Supplier No. PA5111050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QWGNQETSNS GRGRVIQDAE ERPHSRYLRR AYSSDRSGTS NSQSQAKTYT MERCHSAEML SVSKRSGGGE NEERYSPTDN NANIFNFFKE KTSSSSGSFE Highest antigen sequence indentity to the following orthologs: Mouse - 48%, Rat - 51%.

PLK4 or polo-like kinase 4 is a member of the polo family of serine/threonine protein kinases which localizes to centrioles, complex microtubule-based structures found in centrosomes, and regulates centriole duplication during the cell cycle. The overexpression of PLK4 triggered the simultaneous formation of multiple procentrioles around each preexisting centriole which resulting in centriole amplification and thus, Plk4-induced centriole biogenesis in human cells. The reduced Plk4 gene dosage increases the probability of mitotic errors and cancer development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLK4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLK4
Gene Accession No. O00444
Gene Alias 1700028H20; AI385771; MCCRP2; PLK4; PLK-4; polo like kinase 4; polo-like kinase 4; SAK; serine/threonine kinase 18; serine/threonine-protein kinase 18; Serine/threonine-protein kinase PLK4; serine/threonine-protein kinase Sak; Snk akin kinase; Stk18
Gene Symbols PLK4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PLK4. Recombinant protein control fragment (Product #RP-97251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.