Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$192.47 - $517.59

Specifications

Antigen PLK5
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Immunohistochemistry 1:40-1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin 1:40-1:200
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP197306SS
SDP
View Documents
Novus Biologicals
NBP197306SS
0.025 mL
Each for $192.47
Only null left
Add to Cart
 
NBP197306
SDP
View Documents
Novus Biologicals
NBP197306
0.1 mL
Each for $517.59
Only null left
Add to Cart
 
Description

Description

PLK5 Polyclonal specifically detects PLK5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PLK5
Western Blot 1:1000, Immunohistochemistry 1:40-1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin 1:40-1:200
Polyclonal
Rabbit
Neuroscience
Tris-Glycine (pH 7.5) and 0.15M NaCl with 0.05% Sodium Azide
MGC118807, MGC118808, PLK5P, polo-like kinase 5, polo-like kinase 5, pseudogene, SgK384ps
PLK5
IgG
Affinity Purified
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
Q496M5, Q4FZD7
126520
A synthetic peptide made to an internal portion of the human PLK5 protein (between residues 200-250) [UniProt Q496M5]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.