Learn More
Invitrogen™ PLK5 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PLK5 protein.
Specifications
Specifications
| Antigen | PLK5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Plk5 |
| Gene Accession No. | Q496M5 |
| Gene Alias | 6330514A18Rik; FG060302; Inactive serine/threonine-protein kinase PLK5; PLK5; PLK-5; PLK5P; polo like kinase 5; polo-like kinase 5; polo-like kinase 5 pseudogene; polo-like kinase 5, pseudogene; serine/threonine-protein kinase PLK5; SgK384ps |
| Gene Symbols | Plk5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.