Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557120 Supplier Invitrogen™ Supplier No. PA557120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YTVLTGTPPF MASPLSEMYQ NIREGHYPEP AHLSANARRL IVHLLAPNPA ERPSLDHLLQ DDFFTQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.

Polo-like kinase 5 (PLK5) is a member of the Polo-like kinase family, notable for its involvement in cell cycle regulation processes such as mitotic entry, spindle formation, and cytoplasmic division. PLK5 is distinguished from other family members by its low expression levels, particularly within different immunophenotypes, except for being notably elevated in the C5 subtype. Its expression has been linked to poor prognosis in non-small cell lung cancer, although this is not universally supported by all data sources, suggesting complex roles in tumor biology. In a mouse model, PLK5 functions as a DNA damage-inducible gene with nucleolar localization, facilitated by a specific nucleolus localization signal in its N-terminal. The protein's ectopic expression can lead to G1 phase arrest, indicating its role in cellular response to DNA damage. Research is ongoing to fully elucidate its function and prognostic implications across various cancer types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLK5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Plk5
Gene Accession No. Q496M5
Gene Alias 6330514A18Rik; FG060302; Inactive serine/threonine-protein kinase PLK5; PLK5; PLK-5; PLK5P; polo like kinase 5; polo-like kinase 5; polo-like kinase 5 pseudogene; polo-like kinase 5, pseudogene; serine/threonine-protein kinase PLK5; SgK384ps
Gene Symbols Plk5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLK5. Recombinant protein control fragment (Product #RP-97250).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126520
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.