Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLK5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA572987

Catalog No. PIPA572987


Only null left
Add to Cart

Description

Description

PLK5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Inactive serine/threonine-protein kinase that plays a role in cell cycle progression and neuronal differentiation. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLK5
Polyclonal
Unconjugated
Plk5
6330514A18Rik; FG060302; Inactive serine/threonine-protein kinase PLK5; PLK5; PLK-5; PLK5P; polo like kinase 5; polo-like kinase 5; polo-like kinase 5 pseudogene; polo-like kinase 5, pseudogene; serine/threonine-protein kinase PLK5; SgK384ps
Rabbit
Antigen Affinity Chromatography
RUO
126520, 216166
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
TBS with 0.05% sodium azide
Q496M5, Q4FZD7
Plk5
A synthetic peptide made to an internal portion of the human PLK5 protein (between residues 200-250) [UniProt Q496M5].
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.