Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLK5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988123
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8123 Supplier Bioss Supplier No. BS7918R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Inactive serine/threonine-protein kinase that plays a role in cell cycle progression and neuronal differentiation. Ota T., Nat. Genet. 36:40-45(2004). Li H., Submitted (MAR-2006) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004).

Specifications

Antigen PLK5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Plk5
Gene Accession No. Q496M5
Gene Alias 6330514A18Rik; FG060302; Inactive serine/threonine-protein kinase PLK5; Plk5; PLK-5; PLK5P; polo like kinase 5; Polo-like kinase 5; polo-like kinase 5, pseudogene; serine/threonine-protein kinase PLK5; SgK384ps
Gene Symbols Plk5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PLK5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126520
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.