Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLKK Rabbit anti-Human, Clone: JJ080-9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267859
Description
PLKK Monoclonal specifically detects PLKK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
PLKK | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:50, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
PLK, STPK13 | |
Rabbit | |
Protein A purified | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
JJ080-9 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
PLK1 | |
Synthetic phospho-peptide corresponding to residues surrounding Thr210 of Human PLKK aa 181-230/ 603. (SwissProt: P53350 Human) | |
100 μL | |
5347 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction