Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXYD1 Polyclonal Antibody
FXYD1 Polyclonal Antibody
Supplier: Thermo Scientific PA582942
Description
FXYD1 Polyclonal Antibody for ICC/IF

Specifications
FXYD1 | |
Polyclonal | |
Unconjugated | |
FXYD1 | |
0610012C17Rik; 1110006M24Rik; FXYD domain containing ion transport regulator 1; FXYD domain-containing ion transport regulator 1; FXYD1; Phospholemman; PLM; Pml; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00168 | |
FXYD1 | |
Recombinant protein corresponding to Human FXYD1. Recombinant protein control fragment (Product #RP-90196). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction