Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLOD1 Monoclonal Antibody (2114CT109.4.71.73)
GREENER_CHOICE

Catalog No. PIMA537784
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537784 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537784 Supplier Invitrogen™ Supplier No. MA537784
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLOD1 Monoclonal Antibody for Western Blot

Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLOD1
Applications Western Blot
Classification Monoclonal
Clone 2114CT109.4.71.73
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PLOD1
Gene Accession No. Q02809
Gene Alias 2410042F05Rik; AI854890; AV007929; EDS6; LH; LH1; LLH; lysine hydroxylase; lysyl hydroxlase 1; Lysyl hydroxylase 1; Plod; Plod1; procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Symbols PLOD1
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 61-94 amino acids from the C-terminal region of human PLOD1.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.