Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121155 Supplier Invitrogen™ Supplier No. PA5121155
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, A-431, U-251MG, Mouse kidney, Mouse heart. The target is usually found in the following locations: Lumenal side, Peripheral membrane protein, Rough endoplasmic reticulum membrane. Immunogen sequence: KGDAKPEDNL LVLTVATKET EGFRRFKRSA QFFNYKIQAL GLGEDWNVEK GTSAGGGQKV RLLKKALEKH ADKEDLVILF ADSYDVLFAS GPRELLKKFR QARSQVVFSA EELIYPDRRL ETKYPVVSDG KRFLGSGGFI GYAPNLSKLV AEWEGQDSDS DQLFYTKIFL DPEKREQINI TLDHRCRIFQ NLDGALDEVV LKFEMGHVRA RNLAYDTLPV LIHGNGPTKL QLNYLGNYIP RFWTFETGCT VCDEGLRSLK GIGDEALPTV LVGVFIE.

Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLOD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLOD1
Gene Accession No. Q02809, Q63321, Q9R0E2
Gene Alias 2410042F05Rik; AI854890; AV007929; EDS6; LH; LH1; LLH; lysine hydroxylase; lysyl hydroxlase 1; Lysyl hydroxylase 1; Plod; Plod1; procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Symbols PLOD1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116552, 18822, 5351
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.