Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLOD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121676
This item is not returnable.
View return policy
Description
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
Specifications
PLOD1 | |
Polyclonal | |
Unconjugated | |
PLOD1 | |
2410042F05Rik; AI854890; AV007929; EDS6; LH; LH1; LLH; lysine hydroxylase; lysyl hydroxlase 1; Lysyl hydroxylase 1; Plod; Plod1; procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5351 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q02809 | |
PLOD1 | |
Gln636-Pro727 (Accession Q02809), with N-terminal His Tag. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction