Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLOD2 Mouse anti-Human, Clone: 1H9E1, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 663421IG150UL

Description
The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
PLOD2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00469 | |
PLOD2 | |
PLOD2 Fusion Protein Ag5779 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgA |
Immunohistochemistry (Paraffin), Western Blot | |
1H9E1 | |
Unconjugated | |
PLOD2 | |
LH2, Lysyl hydroxylase 2, PLOD2, TLH | |
Mouse | |
Affinity Chromatography | |
RUO | |
5352 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction