Learn More
Invitrogen™ PLOD3 Polyclonal Antibody

Description
PLOD3 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PLOD3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PLOD3 |
| Gene Accession No. | O60568 |
| Gene Alias | AI414586; bone protein I (BP-I); Galactosylhydroxylysine-glucosyltransferase; LH3; lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 2; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; Plod3; Procollagen galactosyltransferase; Procollagen glucosyltransferase; Procollagen glycosyltransferase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; procollagen-lysine,2-oxoglutarate 5-dioxygenase 3 |
| Gene Symbols | PLOD3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.