Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLOD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551546
Description
Immunogen sequence: FDRNRVRIRN VAYDTLPIVV HGNGPTKLQL NYLGNYVPNG WTPEGGCGFC NQDRRTLPGG QPPPRVFLAV FVEQPTPFLP RFLQRLLLLD YPPDRVTLFL HNNEVFHEPH IADSWPQLQD HFSAVKLVGP EEAL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
Specifications
| PLOD3 | |
| Polyclonal | |
| Unconjugated | |
| PLOD3 | |
| AI414586; bone protein I (BP-I); Galactosylhydroxylysine-glucosyltransferase; LH3; lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 2; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; Plod3; Procollagen galactosyltransferase; Procollagen glucosyltransferase; Procollagen glycosyltransferase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; procollagen-lysine,2-oxoglutarate 5-dioxygenase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60568 | |
| PLOD3 | |
| Recombinant protein corresponding to Human PLOD3. Recombinant protein control fragment (Product #RP-94769). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction