Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLP1 Monoclonal Antibody (plpc 1)
GREENER_CHOICE

Catalog No. PIMA190652
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190652 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190652 Supplier Invitrogen™ Supplier No. MA190652
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PLP1 is a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.This gene encodes a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLP1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone plpc 1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene PLP1
Gene Accession No. P04116, P60201
Gene Alias DM20; GPM6C; HLD1; jimpy; jp; lipophilin; lipophilin (aa 129 to 276); major myelin proteolipid protein; MMPL; msd; Myelin proteolipid protein; myelin synthesis deficiency; PLP; PLP/DM20; Plp1; PMD; proteolipid; proteolipid protein; proteolipid protein (myelin) 1; Proteolipid protein (Pelizaeus-Merzbacher disease spastic paraplegia 2 uncomplicated); Proteolipid protein (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein 1; proteolipid protein 1 (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein, lipophilin; rsh; rump shaker; SPG2
Gene Symbols PLP1
Host Species Mouse
Immunogen Synthetic peptide GRGTKF corresponding to C terminal region of myelin proteolipid protein.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281410, 5354
Target Species Human, Bovine, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.