Learn More
Invitrogen™ PLP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA3151
Description
PA3-151 detects proteolipid protein in mouse samples. PA3-151 has been used successfully in Western blot and immunocytochemistry procedures. By Western blot this antibody detects a ~23-26 kDa proteins representing the proteolipid protein and the splice variant DM20 in adult mouse brain samples. The PA3-151 immunogen is a synthetic peptide corresponding to the C-terminal residues K(269) L M G R G T K(276) of PLP.
PLP1 is a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.This gene encodes a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.
Specifications
PLP1 | |
Polyclonal | |
Unconjugated | |
PLP1 | |
DM20; GPM6C; HLD1; jimpy; jp; lipophilin; lipophilin (aa 129 to 276); major myelin proteolipid protein; MMPL; msd; Myelin proteolipid protein; myelin synthesis deficiency; PLP; PLP/DM20; Plp1; PMD; proteolipid; proteolipid protein; proteolipid protein (myelin) 1; Proteolipid protein (Pelizaeus-Merzbacher disease spastic paraplegia 2 uncomplicated); Proteolipid protein (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein 1; proteolipid protein 1 (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein, lipophilin; rsh; rump shaker; SPG2 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
Whole serum with 0.05% sodium azide | |
P60202 | |
PLP1 | |
Synthetic peptide corresponding to the C-terminal residues K(269) L M G R G T K(276) of PLP. | |
RUO | |
18823 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.