Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559973 Supplier Invitrogen™ Supplier No. PA559973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGNHSKIVAG VKAMGAALKH RAKGLRSQGP FLP Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 36%.

PLP2 may play a role in cell differentiation in the intestinal epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLP2
Gene Accession No. Q04941
Gene Alias A4; A4 differentiation-dependent protein; A4LSB; A4-LSB; differentiation-dependent protein A4; differentiation-dependent protein A4; intestinal membrane A4 protein; intestinal membrane A4 protein; mIMA4; PLP2; Proteolipid protein 2; proteolipid protein 2 (colonic epithelium-enriched); Proteolipid protein 2 (intestinal membrane A4 protein)
Gene Symbols PLP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLP2. Recombinant protein control fragment (Product #RP-91118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5355
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.