Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143954
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143954 Supplier Invitrogen™ Supplier No. PA5143954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human COLO320 whole cell, human A431 whole cell. IHC: human adenocarcinoma of the right colon tissue, human cervical cancer tissue, human gastric carcinoma tissue, human liver cancer tissue, human lymphoma tissue, human placenta tissue. Flow: Daudi cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

PLP2 may play a role in cell differentiation in the intestinal epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLP2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene PLP2
Gene Accession No. Q04941
Gene Alias A4; A4 differentiation-dependent protein; A4LSB; A4-LSB; differentiation-dependent protein A4; differentiation-dependent protein A4; intestinal membrane A4 protein; intestinal membrane A4 protein; mIMA4; PLP2; Proteolipid protein 2; proteolipid protein 2 (colonic epithelium-enriched); Proteolipid protein 2 (intestinal membrane A4 protein)
Gene Symbols PLP2
Host Species Rabbit
Immunogen E.coli-derived human PLP2 recombinant protein (Position: N18-V152).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5355
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.