Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542721
Description
Peptide sequence: LVERGNHSKI VAGVLGLIAT CLFGYDAYVT FPVRQPRHTA APTDPADGPV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 93%; Rat: 93%; Sheep: 100%.
PLP2 may play a role in cell differentiation in the intestinal epithelium.
Specifications
PLP2 | |
Polyclonal | |
Unconjugated | |
PLP2 | |
A4; A4 differentiation-dependent protein; A4LSB; A4-LSB; differentiation-dependent protein A4; differentiation-dependent protein A4; intestinal membrane A4 protein; intestinal membrane A4 protein; mIMA4; PLP2; Proteolipid protein 2; proteolipid protein 2 (colonic epithelium-enriched); Proteolipid protein 2 (intestinal membrane A4 protein) | |
Rabbit | |
Protein A | |
RUO | |
5355 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q04941 | |
PLP2 | |
Synthetic peptide directed towards the middle region of human PLP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction