Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLRG1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572038
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2038 10 μL
50-157-2037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2038 Supplier Bethyl Laboratories, Inc Supplier No. IHC00404T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. laevis, X. tropicalis, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, O

This gene encodes a core component of the cell division cycle 5-like complex. The CDC5L complex is part of the spliceosome and is required for pre-mRNA splicing. The encoded protein plays a critical role in alternative splice site selection. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

Specifications

Antigen PLRG1
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PLRG1
Gene Accession No. O43660, Q922V4
Gene Alias PLRG1; pleiotropic regulator 1 (PRL1 homolog, Arabidopsis); PRL1; MGC110980; pleiotropic regulator 1 (PRL1, Arabidopsis homolog); pleiotropic regulator 1 (PRL1homolog, Arabidopsis); pleiotropic regulator 1 (PRL1, Arabidopsis homolog)
Gene Symbols PLRG1
Host Species Rabbit
Immunogen Between 464 and 514
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53317, 5356
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.