Learn More
PLRG1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301940AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
This gene encodes a core component of the cell division cycle 5-like complex. The CDC5L complex is part of the spliceosome and is required for pre-mRNA splicing. The encoded protein plays a critical role in alternative splice site selection. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
PLRG1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O43660, Q922V4 | |
PLRG1 | |
Between 464 and 514 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
PLRG1 | |
PLRG1; pleiotropic regulator 1 (PRL1 homolog, Arabidopsis); PRL1; MGC110980; pleiotropic regulator 1 (PRL1, Arabidopsis homolog); pleiotropic regulator 1 (PRL1homolog, Arabidopsis); pleiotropic regulator 1 (PRL1, Arabidopsis homolog) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53317, 5356 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.