Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590361
Description
Immunogen sequence: MENSTTTISR EELEELQEAF NKIDIDNSGY VSDYELQDLF KEASLPLPGY KVREIVEKIL SVADSNKDGK ISFEEFVSLM QELKSKDISK TFRKIINKRE GITAIGGTST; Positive Samples: HT-29, 293T, LO2, Mouse brain, Mouse liver, Rat spleen; Cellular Location: Cytoplasm.
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. The protein encoded by this gene is a third distinct plastin isoform, which is specifically expressed at high levels in the small intestine. Alternatively spliced transcript variants varying in the 5' UTR, but encoding the same protein, have been found for this gene. A pseudogene of this gene is found on chromosome 11.
Specifications
PLS1 | |
Polyclonal | |
Unconjugated | |
PLS1 | |
AI427122; fimbrin; hypothetical protein LOC334273; I plastin; I-fimbrin; intestine specific plastin; intestine-specific plastin; I-plastin; L-plastin; plastin 1; plastin 1 (I isoform); plastin 1 (I-isoform); plastin 1, I; plastin-1; PLS1; wu:fi38g03; zgc:63494 | |
Rabbit | |
Affinity chromatography | |
RUO | |
102502, 315926, 5357 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.03 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q14651, Q3V0K9 | |
PLS1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human PLS1 (NP_0011657831). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction