Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | PLS1 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PLS1 Polyclonal antibody specifically detects PLS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PLS1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
5357 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
fimbrin, I plastin, intestine specific plastin, Intestine-specific plastin, I-plastin, plastin 1, plastin 1 (I isoform), Plastin-1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DIDNSGYVSDYELQDLFKEAS | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title