Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLS1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571787
Description
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. The protein encoded by this gene is a third distinct plastin isoform, which is specifically expressed at high levels in the small intestine. Alternatively spliced transcript variants varying in the 5′ UTR, but encoding the same protein, have been found for this gene. A pseudogene of this gene is found on chromosome 11.
Specifications
PLS1 | |
Polyclonal | |
Unconjugated | |
PLS1 | |
Plastin-1; Intestine-specific plastin; I-plastin; PLS1 | |
Rabbit | |
Protein A | |
RUO | |
5357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.84 mg/mL | |
PBS with 0.09% sodium azide | |
Q14651 | |
PLS1 | |
KLH conjugated synthetic peptide between 550-579 amino acids from the C-terminal region of human PLS1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction