Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLS3 Monoclonal Antibody (C4), Invitrogen™

Catalog No. FEMA551120
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551120 20 μL
FEMA551119 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551120 Supplier Invitrogen™ Supplier No. MA551120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phospholipid scramblases (PLS) translocate phospholipids bidirectionally between inner and outer leaflets of the plasma membrane and are implicated in apoptosis (1,2). Members of this family, PLS1 and PLS3 are both substrates of Protein kinase C (PKC) delta and are phosphorylated by PKC-delta during apoptosis. PKC-delta translocates to mitochondria during apoptosis and phosphorylates PLS3. Overexpression of PLS3 in the HEK293 cells enhanced apoptosis induced by UV-irradiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLS3
Applications Western Blot
Classification Monoclonal
Clone C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene PLS3
Gene Accession No. P13797, Q63598
Gene Alias AI115446; AL024105; BMND18; LOW QUALITY PROTEIN: plastin-3; MGC128570 protein; plastin 3; plastin 3 (T isoform); plastin 3 (T-isoform); plastin 3, T; plastin-3; PLS3; T fimbrin; T plastin; T-fimbrin; T-plastin
Gene Symbols PLS3
Host Species Mouse
Immunogen Recombinant Human PLS3 protein, Ala379-Val630 (Accession #P13797).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100158187, 5358, 81748
Target Species Human, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.