Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLS3 Monoclonal Antibody (GT3310)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527772
Description
Positive Control: 293T, A431, HeLa Predicted Reactivity: Zebrafish (89%), Xenopus laevis (91%), Chicken (95%), Rhesus Monkey (100%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phospholipid scramblases (PLS) translocate phospholipids bidirectionally between inner and outer leaflets of the plasma membrane and are implicated in apoptosis (1,2). Members of this family, PLS1 and PLS3 are both substrates of Protein kinase C (PKC) delta and are phosphorylated by PKC-delta during apoptosis. PKC-delta translocates to mitochondria during apoptosis and phosphorylates PLS3. Overexpression of PLS3 in the HEK293 cells enhanced apoptosis induced by UV-irradiation.
Specifications
| PLS3 | |
| Monoclonal | |
| 2.39 mg/mL | |
| PBS with no preservative | |
| P13797, Q63598, Q99K51 | |
| PLS3 | |
| Recombinant protein encompassing a sequence within the center region of human T-Plastin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT3310 | |
| Unconjugated | |
| PLS3 | |
| AI115446; AL024105; BMND18; LOW QUALITY PROTEIN: plastin-3; MGC128570 protein; plastin 3; plastin 3 (T isoform); plastin 3 (T-isoform); plastin 3, T; plastin-3; PLS3; T fimbrin; T plastin; T-fimbrin; T-plastin | |
| Mouse | |
| Protein A | |
| RUO | |
| 102866, 5358, 81748 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction